Monitor alcohol and/or ammonia in human serum with bi-level, assayed liquid MAS Alcohol and Ammonia Controls.
Have Questions?
Change view
Catalog Number
Description
Level
10014443
Alcohol Ammonia Controls, Vista Vials, Bi-level, Assayed
Bi-level
AAC-MP
Alcohol and Ammonia Controls, Bi-level, Assayed
Bi-level
Catalog number 10014443
Price (USD)/ Each
-
Request A Quote
Description:
Alcohol Ammonia Controls, Vista Vials, Bi-level, Assayed
Level:
Bi-level
Monitor alcohol and/or ammonia in human serum with bi-level, assayed liquid Thermo Scientific™ MAS™ Alcohol and Ammonia Controls. Available in a convenient Vista™ vial format, this ready-to-use product is packed in convenient multipacks.
Human-serum based matrix
Normal ammonia values in level 1
Assayed values available for most major analyzers
For In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.
Specifications
DescriptionAlcohol Ammonia Controls, Vista Vials, Bi-level, Assayed
Detectable AnalytesAlcohol, Ammonia
Sample TypeSerum
TypeControl
LevelBi-level
Packaging6 Pk.
Quantity2 mL
Unit SizeEach
Contents & Storage
24 month shelf life from date of manufacture at 2° to 8°C; 30 day open vial stability for all analytes at 2° to 8°C