Search Thermo Fisher Scientific
Ensure accurate calibration with the Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Control Set, designed for urine screening with the Thermo Scientific DRI Fentanyl II Drugs of Abuse Assay.
Ensure accurate calibration with the Thermo Scientific DRI Fentanyl II High and Low Control Set, designed for urine screening with the Thermo Scientific DRI Fentanyl II Drug of Abuse Assay. The control set includes a high and low control, targeted at 1.5 ng/mL and 0.5 ng/mL respectively, to validate the calibration curve by providing positive and negative results.