Pacific Hemostasis™ Coagulation Controls
Pacific Hemostasis™ Coagulation Controls
Actual product may vary
Thermo Scientific™
Pacific Hemostasis™ Coagulation Controls
Provide accurate, reproducible confirmation of coagulation test system performance with these coagulation controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
100595TS
also known as 100595
Coagulation Control Level 1
100596TS
also known as 100596
Coagulation Control Level 2
100597TS
also known as 100597
Coagulation Control Level 3
100607TS
also known as 100607
Fibrinogen High Control
100604Fibrinogen Low Control
100013TS1
also known as 100013
Thrombin Time Abnormal Control
Catalog number 100595TS
also known as 100595
Price (USD)
-
Request A Quote
Type:
Coagulation Control Level 1
Provide accurate, reproducible confirmation of coagulation test system performance with Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls. Controls can be run on most coagulation systems and are formulated to reflect expected patient results.
Coagulation and Heparin control plasmas are prepared from fresh pools of human plasma.
Coagulation Control Plasmas:
  • Normal control (Level 1) produces normal PT and aPTT times
  • Abnormal controls monitor therapeutic (Level 2) and highly elevated (Level 3) PT/aPTT times
Heparin Control Plasmas:
  • Simulate low (Level 1) and elevated (Level 2) plasma levels of Heparin
  • Use to monitor and evaluate patients on Heparin therapy
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
TypeCoagulation Control Level 1
Quantity10 x 1 mL
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link