Discover the future of Gibco Media selection with our upcoming updated experience. View details >
Gibco™
KaryoMAX™ Giemsa Stain Solution
Gibco™ KaryoMAX™ Giemsa Stain Solution is a solution used for G-banding of chromosomes for cytogenetic analysis. Gibco™ KaryoMAX™ Giemsa StainRead more
Have Questions?
Catalog number 10092013
Price (USD)/ 100 ml
75.25
-
Add to cart
Price (USD)/ 100 ml
75.25
Add to cart
KaryoMAX™ Giemsa Stain Solution
Catalog number10092013
Price (USD)/ 100 ml
75.25
-
Add to cart
Gibco™ KaryoMAX™ Giemsa Stain Solution is a solution used for G-banding of chromosomes for cytogenetic analysis. Gibco™ KaryoMAX™ Giemsa Stain Solution is formulated as 6.0 g Azur II Eosin and 1.6 g Azur II per liter in glycerol/methanol.
cGMP Manufacturing and Quality System Gibco™ KaryoMAX™ Giemsa Stain Solution is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
FormLiquid
Product TypeGiemsa Stain
Shelf Life24 Months
SterilitySterile-filtered
For Use With (Application)Translational Research, Clinical Research
Product LineKaryoMAX™
Quantity100 mL
Shipping ConditionRoom Temperature
Unit Size100 ml
Contents & Storage
Storage conditions: 15 – 30°C. Protect from light. Shipping conditions: Ambient Shelf life: 24 months from date of manufacture