Informational:This product has been Gibco FBS Fingerprinted. We developed a technology in partnership with Oritain, a global leader in verifying product origins. Based on geographical differences, we can create "fingerprints" of our FBS that connect them to their origin. Find out more ›
Informational:Price on request—please contact your FBS sales representative.
Informational:

***ATTENTION***

*** NEW CATALOG NUMBER***



These products are being replaced with Premium Plus (FBS) performance-based products. The Premium product specifications are improved and product stability, performance, manufacturing, storage, and transport processes are unchanged.

New and improved replacement available, Buy now›
Gibco™

Fetal Bovine Serum, qualified, Australia

Catalog number: 10099141
Have Questions?
Gibco™

Fetal Bovine Serum, qualified, Australia

Catalog number: 10099141
Have Questions?
Catalog Number
10099141
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
10099141500 mL
-
100991581,000 mL
-
A316100150 mL
-
A316100210 x 50 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Premium (Secure)
  • Origin: Australia.
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤30 mg/dL (levels routinely ≤25 mg/dL)

Aliquot-free 50 mL bottle

The Gibco One Shot 50 mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.

Features include:

  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500 mL bottles

All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.

Specifications

Age
Fetal
Species
Cattle/Bovine
Country of Origin
Australia
Form
Liquid
Format
Bottle
Product Type
Fetal Bovine Serum
Quantity
500 mL
Serum Treatment
Standard (Sterile-filtered)
Sterility
Sterile
Endotoxin Concentration
≤5 EU/mL
Green Features
Sustainable packaging
Hemoglobin Concentration
≤30 mg/dL
Purity or Quality Grade
Qualified
Shipping Condition
Frozen

Contents & Storage

Storage conditions: ≤-10°C
Shipping conditions: Frozen

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text