Search Thermo Fisher Scientific
The Thermo Scientific™ MAX-iAQ™ is a fully automated 20-point ambient air monitoring solution for low-level detection of trace compounds, even in high humidity environments. The MAX-iAQ system can quantify 1-100+ compounds down to single digit ppb using advanced Thermo Scientific™ StarBoost™ Technology while maintaining short cycle times. This makes the MAX-iAQ an ideal alternative to traditional gas chromatograph-based systems that are typically slower, less sensitive and more costly to maintain.
Sample Type | Ambient EtO Monitoring |
Voltage | 120 V |
Unit Size | Each |
Sample Type | Ambient Air Monitoring |
Voltage | 120 V |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Sample Type | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
101-0348 | Each | Ambient EtO Monitoring | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
101-0386 | Each | Ambient Air Monitoring | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
MAX-iAQ Ambient Air Monitoring System Overview
The Thermo Scientific™ MAX-iAQ™ is a fully automated 20-point ambient air monitoring solution for low-level detection of trace compounds, even in high humidity environments. The MAX-iAQ system can quantify 1-100+ compounds down to single digit ppb using advanced Thermo Scientific™ StarBoost™ Technology while maintaining short cycle times. This makes the MAX-iAQ an ideal alternative to traditional gas chromatograph-based systems that are typically slower, less sensitive and more costly to maintain.
The MAX-iAQ is designed to operate as a continuous monitor with little to no user interaction, complete with dynamic alarming functionality, user-definable sequencing that can vary by time of day, remote monitoring, and standard data publishing and reporting. Several enclosure options are available depending on specific needs to enable a factory-ready solution.
MAX-iAQ Ambient Air Monitoring System Features