Search Thermo Fisher Scientific
Catalog Number | Product Type |
---|---|
101-0514 | Continuous Emissions Monitoring |
101-0486 | EtO Continuous Emissions Monitor |
The Thermo Scientific™ EMS-10™ Continuous Emissions Monitoring System is a fully automated four-channel stack emission monitoring system designed specifically for hot wet samples. The EMS-10 system can quantify 10-100+ compounds down to single digit ppb while maintaining short cycle times, using advanced FTIR spectral analysis and Thermo Scientific™ StarBoost™ Technology. This makes the EMS-10 an ideal alternative to traditional gas chromatograph-based systems that are typically slower, less sensitive, and more costly to maintain.
EMS-10 FTIR Continuous Emissions Monitoring System Overview
The Thermo Scientific EMS-10 Continuous Emissions Monitoring System is a fully automated four-channel stack emission monitoring system designed specifically for hot wet samples. The EMS-10 system can quantify 10-100+ compounds down to single digit ppb while maintaining short cycle times, using advanced FTIR spectral analysis and Thermo Scientific StarBoost Technology. This makes the EMS-10 an ideal alternative to traditional gas chromatograph-based systems that are typically slower, less sensitive, and more costly to maintain.
The EMS-10 is designed to operate as a continuous emissions monitor with little to no user interaction, complete with dynamic alarming functionality, user-definable sequencing that can vary by time of day, remote monitoring, and standard data publishing and reporting. Several enclosure options are available depending on specific needs to enable a factory-ready solution.
The integrated design incorporates complete control of all gases, including the sample stream, zero gas and calibration gas, with software from the large front panel touchscreen or keyboard. This system is ideal whenever and wherever gas streams must be analyzed for composition, impurities, and hazardous air pollutants.
EMS-10 Continuous Emission Monitoring System Features
Continuous Emission Monitoring System Uses
Recently approved US EPA regulations address air quality and point source emissions from industries that use ethylene oxide (EtO). Companies must prepare to adopt new equipment and controls for monitoring EtO in compliance with updated NESHAP guidelines. OE-FTIR spectroscopy stands out for its ability to analyze EtO emissions at concentrations as low as 1 ppb, even in challenging sample matrices off a stack. Learn how OE-FTIR technology can be used to provide high-quality data, even amid interferences and sample variability, and in turn help facilities that use EtO achieve compliance with new stricter standards.
Share catalog number, name or link