Orion™ Star Series RS232 to USB Cable Adapter
Product Image
Thermo Scientific™
Orion™ Star Series RS232 to USB Cable Adapter
Connect a USB port to the RS232 port of your meter using the Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter.
Have Questions?
Catalog NumberForUseWithEquipment
1010005
Catalog number 1010005
Price (USD)
155.70
Online Exclusive
173.00
Save 17.30 (10%)
Each
-
Add to cart
Price (USD)
155.70
Online Exclusive
173.00
Save 17.30 (10%)
Each
Add to cart
Ask our AI about this Product
Connect a USB port to the RS232 port of your meter using the Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter. This adapter allows interface between a USB connection and Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series (Star A320, Star A220 and Star A210 series) meters and legacy Orion™ Star and Star Plus meters.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionOrion Star series RS232 to USB adapter cable
IncludesRS232 to USB computer interface adapter and cable (2.5mm RS232 meter connector and USB computer connector)
TypeMeter Interface Cable
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection