1x1 image pixel for data collection
Protein A–FITC Conjugate
Protein A–FITC Conjugate
Invitrogen™
Protein A–FITC Conjugate
Protein A (from Staphylococcus aureus, Cowan I strain) is conjugated with fluorescein isothiocyanate (FITC) and is purified by molecular sievingRead more
Have Questions?
Catalog NumberQuantity
1010112 mL
Catalog number 101011
Price (USD)
183.65
Online Exclusive
245.00
Save 61.35 (25%)
Each
In stock
Add to cart
Quantity:
2 mL
Price (USD)
183.65
Online Exclusive
245.00
Save 61.35 (25%)
Each
Add to cart
Protein A (from Staphylococcus aureus, Cowan I strain) is conjugated with fluorescein isothiocyanate (FITC) and is purified by molecular sieving to remove the unbound FITC and unconjugated Protein A. The product is in 10 mM phosphate buffered saline, pH 7.4, containing 1% ovalbumin, 40% glycerol, and 0.1% sodium azide.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormSolution
Protein SubtypeProtein A
Shipping ConditionWet Ice
ConjugateFluorescein
Quantity2 mL
Unit SizeEach
Contents & Storage
Store at 2-8°C
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link