This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A toRead more
Have Questions?
Change view
Catalog Number
Quantity
101042
25 mL
101041
5 mL
101090
also known as 10-1090
1 L
3 Options
Catalog number 101042
Price (USD)
1,018.65
Online Exclusive
1,090.00
Save 71.35 (7%)
Each
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)
1,018.65
Online Exclusive
1,090.00
Save 71.35 (7%)
Each
Add to cart
This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A to Sepharose™ 4B.
This product is supplied as a 50% suspension consisting of 25ml Protein A-Sepharose™ 4B and 25 ml of PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.19% Kathon CG/ICP).