Invitrogen™

Recombinant Protein A-Sepharose™ 4B

Catalog number: 101141
Have Questions?
Invitrogen™

Recombinant Protein A-Sepharose™ 4B

Catalog number: 101141
Have Questions?
Catalog Number
101141
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1011415 mL
-
10114225 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This product is prepared by covalently coupling highly purified Staph. aureus Cowan strain Protein A produced in E. Coli to Sepharose™ 4B.

This product is supplied as a 50% suspension consisting of 25ml recombinant Protein A-Sepharose™ 4B and 25 ml of PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.19% Kathon CG/ICP).

Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Protein A Sepharose
Column Type
Affinity
Stationary Phase
Protein A
Quantity
5 mL
Form
Suspension

Contents & Storage

2°C to 8°C. Do Not Freeze.
This product is supplied as a 50% suspension (5 ml Recombinant Protein A-Sepharose™ 4B + 5 ml PBS) Wet Ice

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text