Invitrogen™

Recombinant Protein G

Catalog number: 101200
Have Questions?
Invitrogen™

Recombinant Protein G

Catalog number: 101200
Have Questions?
Catalog Number
101200
Unit Size
1 MG
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 MG
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1012001 MG
-
1012015 MG
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. It is supplied as a salt-free lyophilized powder.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

• all four subclasses of human IgG
• mouse IgG
• rabbit IgG
• guinea pig IgG
• rat IgG
• goat IgG
• sheep IgG
• cow IgG
• horse IgG

Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Conjugate
Unconjugated
Quantity
1 mg
Form
Powder
Protein Subtype
Protein G
Shipping Condition
Wet Ice

Contents & Storage

2-8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text