Invitrogen™

Recombinant Protein G-HRP

Catalog number: 101223
Have Questions?
Invitrogen™

Recombinant Protein G-HRP

Catalog number: 101223
Have Questions?
Catalog Number
101223
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1012231 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This protein G is conjugated with highly purified horseradish peroxidase (RZ > 3.0) and is in enyme conjugate stabilizer containing 0.19% Kathon CG/ICP.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

• all four subclasses of human IgG
• mouse IgG
• rabbit IgG
• guinea pig IgG
• rat IgG
• goat IgG
• sheep IgG
• cow IgG
• horse IgG

Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Conjugate
HRP
Quantity
1 mL
Form
Powder
Protein Subtype
Protein G
Shipping Condition
Wet Ice

Contents & Storage

2-8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text