Invitrogen™

Recombinant Protein G - Sepharose™ 4B

Catalog number: 101242
Have Questions?
Invitrogen™

Recombinant Protein G - Sepharose™ 4B

Catalog number: 101242
Have Questions?
Catalog Number
101242
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1012425 mL
-
1012412 mL
-
10124325 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This product is prepared by covalently coupling highly purified rec-Protein G to Sepharose™ 4B. It is supplied as a 50% suspension containing 25 ml rec-Protein G-Sepharose™ 4B + 25 ml PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.19% Kathon CG/ICP).

Applications: Immunoglobulin affinity purification and Immunoprecipitation.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

• all four subclasses of human IgG
• mouse IgG
• rabbit IgG
• guinea pig IgG
• rat IgG
• goat IgG
• sheep IgG
• cow IgG
• horse IgG

Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Protein G Sepharose
Product Line
Sepharose™
Column Type
Affinity
Stationary Phase
Protein G
Quantity
5 mL
Form
Suspension

Contents & Storage

This product is supplied as a 50% suspension (5 ml Recombinant Protein G-Sepharose™ 4B + 5 ml PBS) Wet Ice

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text