Geneticin™ Selective Antibiotic (G418 Sulfate) (50 mg/mL)
Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
Geneticin™ Selective Antibiotic (G418 Sulfate) (50 mg/mL)
Gibco™

Geneticin™ Selective Antibiotic (G418 Sulfate) (50 mg/mL)

Gibco Geneticin Selective Antibiotic (G418 Sulfate) is produced by the bacterium Micromonospora rhodorangea and acts by binding the ribosome, thusRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
10131027100 mL
1013103520 mL
Catalog number 10131027
Price (USD)
736.00
Each
-
Add to cart
Quantity:
100 mL
Recurring order eligible. Learn more »
Price (USD)
736.00
Each
Add to cart
Ask our AI about this Product
Gibco Geneticin Selective Antibiotic (G418 Sulfate) is produced by the bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells. Resistance to Geneticin Selective Antibiotic (G418 Sulfate) is conferred by the E. coli APH (3’) – I and APH (3’) – II resistance genes. Geneticin Selective Antibiotic (G418 Sulfate) is used as a selective antibiotic in the concentration range of 100–200 μg/mL for bacteria, or 200–500 μg/mL for most mammalian cells. This product is supplied as a 50 mg/mL solution in water.

Dual-Site cGMP Manufacturing and Quality Standard
For supply chain continuity, we manufacture Gibco Geneticin Selective Antibiotic (G418 Sulfate) at two separate facilities located in Grand Island, NY, and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeEukaryotic Cells, Prokaryotic Cells
Concentration50 mg/mL
Culture TypeMammalian Cell Culture, Insect Cell Culture
For Use With (Application)Eukaryotic Selection/Stable Cell Line Generation
Product LineGeneticin™
Quantity100 mL
Shelf Life24 Months
Shipping ConditionRoom Temperature
FormLiquid
Product TypeAntibiotic
SterilitySterile-filtered
Sterilization MethodSterile-filtered
Unit SizeEach
Contents & Storage
Storage conditions: 2 to 8°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3150452Certificate of AnalysisMay 24, 202510131027, 10131035
3118120Certificate of AnalysisMay 22, 202510131027, 10131035
3126859Certificate of AnalysisMay 07, 202510131027, 10131035
3108042Certificate of AnalysisMar 30, 202510131027, 10131035
2954097Certificate of AnalysisMar 13, 202510131027, 10131035
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

All of our antibiotics (Geneticin, Zeocin, Hygromycin B, Blasticidin, and Puromycin) can be used together for making multiple stable cell lines. However, kill curves will need to be performed for each combination of antibiotics since sensitivity to a given antibiotic tends to increase when combined with other antibiotics.

No, Neomycin is toxic to mammalian cells. It also causes irreversible damage to kidneys and other organs. Geneticin (aka G418 Sulfate) is a less toxic and very effective alternative for selection in mammalian cells.  Neomycin can be used in bacterial selection, but Kanamycin is the preferred drug to use because of Neomycin's toxicity.

For best results, optimal concentrations for selection should be determined empirically in each unique experiment through dose response curves. However, to get a general idea of concentrations that have worked for individual cell types, please click on the following url: http://www.thermofisher.com/us/en/home/life-science/cell-culture/transfection/selection.html or type in “Selection Antibiotics” into our main search on www.thermofisher.com.

It is true that a percentage of non-resistant mammalian cells do not round-up from the plate upon Zeocin selection as would be seen with G418 or Hygromycin selection. However, one should see some very characteristic morphological changes occurring in those cells that are not resistant. These cells that stick to the culture dish typically display a vast increase in size. This could be best described as being similar to the effects of cytomegalovirus infecting permissive cells. The shape of these cells may also change; taking on an "alien" shape. On close examination of the non-resistant cells, the researcher should observe a distinct breakdown of both the nuclear and plasma membranes. Even though the "cells" are still attached to the plate, they should have the appearance of many holes in these membranes. Also, before the breakdown of the membranes, one can observe open areas in the cytoplasm of the cells that appear to be large, empty vesicles. Although not confirmed, this may be explained by a breakdown of the endoplasmic reticulum and Golgi apparatus, or other scaffolding proteins. Eventually, these "cells" will completely breakdown so that only "strings" of protein are left.

In contrast, Zeocin resistant cells should continue to divide at a regular interval to form distinct clumps of cells, or colonies. There should not be a distinct change in morphology, which can be compared to cells not under selection with Zeocin. It is these colonies of actively dividing cells that contain the resistance gene and are expressing it actively.

If there is concern about the dead cells sticking to the plate, one may do the following to eliminate them: Treat the plate for a couple of minutes with trypsin/versene. Both the healthy resistant cells and the dead cells will dislodge from the plate. The cells can then be replated (without Zeocin selection) and the healthy cells will attach again while the dead ones will not. After a couple of hours when the healthy cells have attached to the substrate again, Zeocin can be added back to the medium.

Blasticidin: Nucleoside Inhibits protein synthesis in prokaryotic and eukaryotic cells by interfering with peptidyl transfer reaction of protein synthesis, causing early termination of translation.

Geneticin (G418): Aminoglycoside Blocks protein synthesis in mammalian cells by interfering with ribosomal function.

Hygromycin: Aminocyclitol Inhibits protein synthesis by disrupting translocation and promoting mistranslation.

Zeocin: Intercalates with DNA and cleaves it.

Citations & References (117)

Citations & References
Abstract
Isolation and characterization of a novel cDNA encoding a human UDP-glucuronosyltransferase active on C19 steroids.
Authors:Beaulieu M,Lévesque E,Hum DW,Bélanger A
Journal:The Journal of biological chemistry
PubMed ID:8798464
Cloning and functional expression of a thyrotropin receptor cDNA from rat fat cells.
Authors:Endo T; Ohta K; Haraguchi K; Onaya T;
Journal:J Biol Chem
PubMed ID:7738021
Thyrotropin receptor (TSH-R) has been thought to be thyroid-specific, but, by Northern blot analysis, we found that rat adipose tissue expressed TSH-R mRNAs in amounts approaching those in the thyroid. To investigate the function of TSH-R from adipose tissue, we screened a rat fat cell lambda gt11 cDNA library for ... More
Molecular cloning and functional expression of murine JE (monocyte chemoattractant protein 1) and murine macrophage inflammatory protein 1alpha receptors: evidence for two closely linked C-C chemokine receptors on chromosome 9.
Authors: Boring L; Gosling J; Monteclaro F S; Lusis A J; Tsou C L; Charo I F;
Journal:J Biol Chem
PubMed ID:8631787
'We have isolated cDNA clones that encode two closely related, murine C-C chemokine receptors. Both receptors are members of the G-protein-coupled, seven-transmembrane domain family of receptors and are most closely related to the human monocyte chemoattractant protein 1 receptor. Expression of each of the receptors was detected in murine monocyte/macrophage ... More
Residues throughout the cytoplasmic domain affect the internalization efficiency of P-selectin.
Authors: Setiadi H; Disdier M; Green S A; Canfield W M; McEver R P;
Journal:J Biol Chem
PubMed ID:7592923
'The cytoplasmic domains of many membrane proteins have short sequences, usually including a tyrosine or a di-leucine, that function as sorting signals. P-selectin is an adhesion receptor for leukocytes that is expressed on activated platelets and endothelial cells. Its 35-residue cytoplasmic domain contains signals for sorting into regulated secretory granules, ... More
Structural basis of G protein specificity of human endothelin receptors. A study with endothelinA/B chimeras.
Authors: Takagi Y; Ninomiya H; Sakamoto A; Miwa S; Masaki T;
Journal:J Biol Chem
PubMed ID:7730310
'The endothelin (ET) family of peptides acts via two subtypes of guanine nucleotide-binding regulatory protein (G protein)-coupled receptors termed ETA and ETB. ET-1 stimulated cAMP formation in Chinese hamster ovary (CHO) cells stably expressing human wild-type ETA (CHO/hETA cells) while it inhibited cAMP formation in CHO cells expressing human wild-type ... More
117 total citations

Share catalog number, name or link

1x1 image pixel for data collection