Ramsey™ Mercury-Free Tilt Sensor
Ramsey™ Mercury-Free Tilt Sensor
Thermo Scientific™

Ramsey™ Mercury-Free Tilt Sensor

Prevent jitter and provide stability in your hazardous area applications with the Thermo Scienitifc™ Ramsey™ Mercury-Free Tilt Sensor.

Have Questions?
Catalog NumberProduct TypeTemperature Range
10138045Mercury-Free Tilt SensorControllers: -40° to +50°C (-40° to +122°F)
Catalog number 10138045
Price (USD)
-
Product Type:
Mercury-Free Tilt Sensor
Temperature Range:
Controllers: -40° to +50°C (-40° to +122°F)
Ask our AI about this Product

This product has been discontinued – effective June 26, 2023. Aftermarket support including remote technical support and some field service will continue to be available as well as spare parts based on material availability.

Please check for more details on the discontinuation and contact our service team here.

At Thermo Fisher Scientific, we provide our valued customers with the products, solutions, and support that enable you to make the world healthier, cleaner, and safer.

To ensure we can continue to offer innovation, quality, value, and acceptable lead times we continuously evaluate our portfolio and take the tough decisions to discontinue products where necessary.

Specifications
Product TypeMercury-Free Tilt Sensor
Environmental RatingTilt Sensor: IP67 (NEMA-4X), Controllers: Model 20-35-NM-DIN: IP20 (NEMA 1), All other models: IP65 (NEMA 4 or NEMA 4X)
Alarm Contact RatingControllers: 20-35-NM-DIN: (1) NO—2 Amp @ 115/230VAC, 3 Amp @ 24VDC; All other models: (2) SPDT—10 Amp @ 115/230VAC, 7 Amp @ 30VDC non-inductive
Certifications/ComplianceTilt Sensor and Controllers: All models: cCSAus, CE
Input PowerControllers: 115/230 V, 50/60 Hz
MountingHanger and two S-hooks
Temperature RangeControllers: -40° to +50°C (-40° to +122°F)
Time DelayControllers: 1, 2, 4 or 6 seconds selectable by jumpers
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection