Thermo Scientific™

SGD-O Density Gauge for Fracturing and Cementing

Catalog number: 10139861
Have Questions?
Thermo Scientific™

SGD-O Density Gauge for Fracturing and Cementing

Catalog number: 10139861
Have Questions?
This product has been discontinued; this includes the discontinuation of spare/replacement parts and all service and support.
 
Catalog Number
10139861
Unit Size
Each
Product Type
Density Gauge
Price (USD)
Full specifications
Product TypeDensity Gauge
For Use With (Application)Fracking blending trucks (low pressure) and wellhead (high pressure)
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeProduct TypePrice (USD)
10139861Full specifications
EachDensity Gauge
Product TypeDensity Gauge
For Use With (Application)Fracking blending trucks (low pressure) and wellhead (high pressure)
Unit SizeEach
Showing 1 of 1
This product has been discontinued; this includes the discontinuation of spare/replacement parts and all service and support.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text