Search Thermo Fisher Scientific
Type | Spectrophotometer Test Tube and Special Application Cell |
Description | Test Tube and Special Application Cells |
Wavelength Range | For visible measurements of range 320-1100nm |
For Use With (Application) | Thermo Scientific Test Tube and Special Application Cells for quick measurements that do not require transfer of solution to cuvette, glass test tube cells. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
10145335 | Each | Test Tube and Special Application Cells | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|