Flow Cells
Product Image
Thermo Scientific™
Flow Cells
Thermo Scientific™ Flow Cells are available for use with autosamplers and sipper systems in a variety of pathlengths.
Have Questions?
Catalog NumberDescriptionMaterial
10145336Flow CellsGlass, Quartz
Catalog number 10145336
Price (USD)
-
Request A Quote
Description:
Flow Cells
Material:
Glass, Quartz
Thermo Scientific™ Flow Cells are available for use with autosamplers and sipper systems in a variety of pathlengths. Sloping roof flow cells keep bubbles out of light beam improving quality of data using automated methods.
  • Allows liquid samples to flow through measurement region for automated analysis with an autosampler or a sipper system
  • Glass cells allow visible measurements in 320-1100nm range
  • Quartz cells allow UV-Visible measurements in 190-1100nm range
Use single cell holder for best results.
Specifications
TypeFlow Cell
DescriptionFlow Cells
MaterialGlass, Quartz
Wavelength RangeGlass cells allow visible measurements in 320-1100nm range; Quartz cells allow UV-Visible measuremen
For Use With (Equipment)Thermo Scientific Flow Cells are Available For Use With Autosamplers and Sipper Systems in a Variety of Pathlength
For Use With (Application)Thermo Scientific Flow Cells are available for use with autosamplers and sipper systems in a variety of pathlengths
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link