Acetal, 99%, Thermo Scientific Chemicals
Acetal, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetal, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 102220050
Price (USD)/ Each
25.65
Online exclusive
30.30 
Save 4.65 (15%)
-
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
30.30 
Save 4.65 (15%)
Add to cart
Acetal, 99%, Thermo Scientific Chemicals
Catalog number102220050
Price (USD)/ Each
25.65
Online exclusive
30.30 
Save 4.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,1-diethoxyethane
Molecular FormulaC6H14O2
InChI KeyDHKHKXVYLBGOIT-UHFFFAOYSA-N
SMILESCCOC(C)OCC
Molecular Weight (g/mol)118.18
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Density(20°C) 0.822 to 0.829 kg/l
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only