2-Acetamidophenol, 97%, Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Acetamidophenol, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number 102311000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS614-80-2
IUPAC NameN-(2-hydroxyphenyl)acetamide
Molecular FormulaC8H9NO2
InChI KeyADVGKWPZRIDURE-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point205°C to 210°C
Appearance (Form)Powder
HPLC>=96.0 %
Appearance (Color)Off-white to beige to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only