Acetone cyanohydrin, 99%, stabilized, Thermo Scientific Chemicals
Acetone cyanohydrin, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetone cyanohydrin, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 102380050
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.90 
Save 8.25 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.90 
Save 8.25 (14%)
Add to cart
Acetone cyanohydrin, 99%, stabilized, Thermo Scientific Chemicals
Catalog number102380050
Price (USD)/ Each
50.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
58.90 
Save 8.25 (14%)
-
Add to cart
Chemical Identifiers
CAS75-86-5
IUPAC Name2-hydroxy-2-methylpropanenitrile
Molecular FormulaC4H7NO
InChI KeyMWFMGBPGAXYFAR-UHFFFAOYSA-N
SMILESCC(C)(O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
StabilizerApprox. 0.3% H3PO4
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only