Thermo Scientific Chemicals

Acetone oxime, 98%, Thermo Scientific Chemicals

Catalog number: 102391000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Acetone oxime, 98%, Thermo Scientific Chemicals

Catalog number: 102391000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 102391000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-06-0
IUPAC Name
N-(propan-2-ylidene)hydroxylamine
Molecular Formula
C3H7NO
InChI Key
PXAJQJMDEXJWFB-UHFFFAOYSA-N
SMILES
CC(C)=NO
Infrared spectrum
Conforms
Appearance (Color)
White to yellow
GC
>=97.5 %
Appearance (Form)
Crystals or crystalline solid
Melting point
58°C to 63°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text