Acetovanillone, 98%, Thermo Scientific Chemicals
Acetovanillone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetovanillone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 102420250
Price (USD)/ Each
34.60
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
34.60
Add to cart
Acetovanillone, 98%, Thermo Scientific Chemicals
Catalog number102420250
Price (USD)/ Each
34.60
-
Add to cart
Chemical Identifiers
IUPAC Name1-(4-hydroxy-3-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O3
InChI KeyDFYRUELUNQRZTB-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1O)C(C)=O
Molecular Weight (g/mol)166.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point112°C to 116°C
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only