9-Acetylanthracene, 95%, Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9-Acetylanthracene, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 102440250
Price (USD)/ Each
138.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
138.00
Add to cart
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
Catalog number102440250
Price (USD)/ Each
138.00
-
Add to cart
Chemical Identifiers
IUPAC Name1-(anthracen-9-yl)ethan-1-one
Molecular FormulaC16H12O
InChI KeyNXXNVJDXUHMAHU-UHFFFAOYSA-N
SMILESCC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
Molecular Weight (g/mol)220.27
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=94 %
Appearance (Color)Yellow to yellow-brown
Melting point72°C to 76°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only