2-Acetylbenzoic acid, 99%, Thermo Scientific Chemicals
2-Acetylbenzoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Acetylbenzoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 102470250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-acetylbenzoic acid
Molecular FormulaC9H8O3
InChI KeyQDAWXRKTSATEOP-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1C(O)=O
Molecular Weight (g/mol)164.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Appearance (Color)Light yellow to light orange
Melting point115°C to 118°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only