Thermo Scientific Chemicals

Acetylenedicarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 102521000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Acetylenedicarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 102521000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 102521000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-45-0
IUPAC Name
but-2-ynedioic acid
Molecular Formula
C4H2O4
InChI Key
YTIVTFGABIZHHX-UHFFFAOYSA-N
SMILES
OC(=O)C#CC(O)=O
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
>=97.5 %
Appearance (Color)
White to cream to beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text