4-Acetylpyridine, 98%, Thermo Scientific Chemicals
4-Acetylpyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetylpyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 102700100
Price (USD)/ Each
22.20
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
22.20
Add to cart
4-Acetylpyridine, 98%, Thermo Scientific Chemicals
Catalog number102700100
Price (USD)/ Each
22.20
-
Add to cart
Chemical Identifiers
CAS1122-54-9
IUPAC Name1-(pyridin-4-yl)ethan-1-one
Molecular FormulaC7H7NO
InChI KeyWMQUKDQWMMOHSA-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5270 to 1.5310 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear light yellow to brown
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only