Thermo Scientific Chemicals

Adenosine 5'-monophosphate, 99%, Thermo Scientific Chemicals

Catalog number: 102791000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Adenosine 5'-monophosphate, 99%, Thermo Scientific Chemicals

Catalog number: 102791000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 102791000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
61-19-8
IUPAC Name
{[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Molecular Formula
C10H14N5O7P
InChI Key
UDMBCSSLTHHNCD-YPLCUDRINA-N
SMILES
NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Appearance (Form)
Crystalline powder
UV transmittance
>=95.0 % (c=1, 0.2 N NaOH, 430 nm) 1 cm cell
Appearance (Color)
Colorless to white
Infrared spectrum
Conforms
Loss on drying
=<6 % (105°C, 5 hrs)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text