Thermo Scientific Chemicals

Adipic acid, 99%, Thermo Scientific Chemicals

Catalog number: 102810010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Adipic acid, 99%, Thermo Scientific Chemicals

Catalog number: 102810010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 102810010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-04-9
IUPAC Name
hexanedioic acid
Molecular Formula
C6H10O4
InChI Key
WNLRTRBMVRJNCN-UHFFFAOYSA-N
SMILES
OC(=O)CCCCC(O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
151°C to 153°C
Water
=<0.2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text