Thermo Scientific Chemicals

Monomethyl adipate, 97%, Thermo Scientific Chemicals

Catalog number: 102821000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Monomethyl adipate, 97%, Thermo Scientific Chemicals

Catalog number: 102821000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 102821000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
627-91-8
IUPAC Name
6-methoxy-6-oxohexanoic acid
Molecular Formula
C7H12O4
InChI Key
UOBSVARXACCLLH-UHFFFAOYSA-N
SMILES
COC(=O)CCCCC(O)=O
Appearance (Color)
Clear colorless to light yellow
Titration with NaOH
>=96.0 %
Appearance (Form)
Liquid
Refractive index
1.4390 to 1.4430 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text