Thermo Scientific Chemicals

L-Alanine, 99%, Thermo Scientific Chemicals

Catalog number: 102830250
25 g, Plastic Bottle, Each
Thermo Scientific Chemicals

L-Alanine, 99%, Thermo Scientific Chemicals

Catalog number: 102830250
25 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 102830250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-41-7
IUPAC Name
(2S)-2-aminopropanoic acid
Molecular Formula
C3H7NO2
InChI Key
QNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILES
C[C@H](N)C(O)=O
Chloride (Cl)
=<200 ppm
Sulfate (SO4)
=<300 ppm
Appearance (Color)
White to off-white
pH
5.5 to 7 (2.5 % at 25°C)
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text