L-alanina, 99 %, Thermo Scientific Chemicals
L-alanina, 99 %, Thermo Scientific Chemicals
L-alanina, 99 %, Thermo Scientific Chemicals
L-alanina, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-alanina, 99 %, Thermo Scientific Chemicals

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
25 g
100 g
Envase:
Frasco de plástico
Número de catálogo 102831000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS56-41-7
IUPAC Name(2S)-2-aminopropanoic acid
Molecular FormulaC3H7NO2
InChI KeyQNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILESC[C@H](N)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Chloride (Cl)=<200 ppm
Sulfate (SO4)=<300 ppm
Appearance (Color)White to off-white
pH5.5 to 7 (2.5 % at 25°C)
Appearance (Form)Crystalline powder or crystals
Ver más
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

Hojas de datos de seguridad

Compartir número de catálogo, nombre o enlace.