L-Alanine, 99%
L-Alanine, 99%
L-Alanine, 99%
Thermo Scientific Chemicals

L-Alanine, 99%

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
Catalog number 102835000
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS56-41-7
IUPAC Name(2S)-2-aminopropanoic acid
Molecular FormulaC3H7NO2
InChI KeyQNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILESC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<200 ppm
Sulfate (SO4)=<300 ppm
Appearance (Color)White to off-white
pH5.5 to 7 (2.5 % at 25°C)
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection