Thermo Scientific Chemicals

Allantoin, 98%, Thermo Scientific Chemicals

Catalog number: 102841000
100 g, Each
Thermo Scientific Chemicals

Allantoin, 98%, Thermo Scientific Chemicals

Catalog number: 102841000
100 g, Each
Quantity
Catalog number: 102841000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
97-59-6
IUPAC Name
(2,5-dioxoimidazolidin-4-yl)urea
Molecular Formula
C4H6N4O3
InChI Key
POJWUDADGALRAB-UHFFFAOYNA-N
SMILES
NC(=O)NC1NC(=O)NC1=O
Appearance (Color)
White
Titration with NaOH
>=97.5 %
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text