Thermo Scientific Chemicals

4-Allylanisole, 98%, Thermo Scientific Chemicals

Catalog number: 102880250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Allylanisole, 98%, Thermo Scientific Chemicals

Catalog number: 102880250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 102880250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-67-0
IUPAC Name
1-methoxy-4-(prop-2-en-1-yl)benzene
Molecular Formula
C10H12O
InChI Key
ZFMSMUAANRJZFM-UHFFFAOYSA-N
SMILES
COC1=CC=C(CC=C)C=C1
Refractive index
1.5190 to 1.5240 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text