Allyl Glycidyl Ether, 99+%
Allyl Glycidyl Ether, 99+%
Allyl Glycidyl Ether, 99+%
Allyl Glycidyl Ether, 99+%
Thermo Scientific Chemicals

Allyl Glycidyl Ether, 99+%

CAS: 106-92-3 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 102941000
Price (EUR)
25,76
Online Exclusive
30,30
Save 4,54 (15%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
25,76
Online Exclusive
30,30
Save 4,54 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name2-[(prop-2-en-1-yloxy)methyl]oxirane
Molecular FormulaC6H10O2
InChI KeyLSWYGACWGAICNM-UHFFFAOYNA-N
SMILESC=CCOCC1CO1
Molecular Weight (g/mol)114.14
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4332 to 1.4352 (20°C, 589 nm)
Appearance (Color)Clear colorless
Water=<0.2 % (K.F.)
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link