Allyl Glycidyl Ether, 99+%, Thermo Scientific Chemicals
Allyl Glycidyl Ether, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allyl Glycidyl Ether, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number 102945000
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
Add to cart
Allyl Glycidyl Ether, 99+%, Thermo Scientific Chemicals
Catalog number102945000
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-[(prop-2-en-1-yloxy)methyl]oxirane
Molecular FormulaC6H10O2
InChI KeyLSWYGACWGAICNM-UHFFFAOYNA-N
SMILESC=CCOCC1CO1
Molecular Weight (g/mol)114.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water=<0.2 % (K.F.)
Refractive index1.4332 to 1.4352 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only