Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol, Thermo Scientific Chemicals
Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 102955000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-isothiocyanatoprop-1-ene
Molecular FormulaC4H5NS
InChI KeyZOJBYZNEUISWFT-UHFFFAOYSA-N
SMILESC=CCN=C=S
Molecular Weight (g/mol)99.15
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5300 to 1.5330 (20°C, 589 nm)
Stabilizer0.01 % alpha-Tocopherol
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only