Thermo Scientific Chemicals

Allyl sulfide, 98%, Thermo Scientific Chemicals

Catalog number: 103021000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Allyl sulfide, 98%, Thermo Scientific Chemicals

Catalog number: 103021000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 103021000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
592-88-1
IUPAC Name
3-(prop-2-en-1-ylsulfanyl)prop-1-ene
Molecular Formula
C6H10S
InChI Key
UBJVUCKUDDKUJF-UHFFFAOYSA-N
SMILES
C=CCSCC=C
Appearance (Color)
Clear colorless
Refractive index
1.4879 to 1.4899 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text