Chemical Identifiers
IUPAC Nametriammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular FormulaC22H25N3O9
InChI KeyUDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
Molecular Weight (g/mol)475.45
View more