Thermo Scientific Chemicals

Aluminon, Thermo Scientific Chemicals

Catalog number: 103061000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Aluminon, Thermo Scientific Chemicals

Catalog number: 103061000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 103061000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
569-58-4
IUPAC Name
triammonium 5-[(3-carboxylato-4-hydroxyphenyl)(3-carboxylato-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoate
Molecular Formula
C22H25N3O9
InChI Key
UDPQMNAHKKTHHI-UHFFFAOYNA-N
SMILES
[NH4+].[NH4+].[NH4+].OC1=CC=C(C=C1C([O-])=O)C(C1C=CC(=O)C(=C1)C([O-])=O)C1=CC=C(O)C(=C1)C([O-])=O
Appearance (Form)
Crystalline powder
Sulfated ash
=<0.2 %
Appearance (Color)
Red to yellow-brown to red-brown
Infrared spectrum
Conforms
Suitability
(for Al determination) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text