Thermo Scientific Chemicals

Aminoacetaldehyde Diethyl Acetal, 99%, Thermo Scientific Chemicals

Catalog number: 103070500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

Aminoacetaldehyde Diethyl Acetal, 99%, Thermo Scientific Chemicals

Catalog number: 103070500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 103070500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
645-36-3
IUPAC Name
2,2-diethoxyethan-1-amine
Molecular Formula
C6H15NO2
InChI Key
HJKLEAOXCZIMPI-UHFFFAOYSA-N
SMILES
CCOC(CN)OCC
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4160 to 1.4180 (20°C, 589 nm)
GC
>=98.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text