4-Aminoantipyrine, 98%, Thermo Scientific Chemicals
4-Aminoantipyrine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminoantipyrine, 98%, Thermo Scientific Chemicals

4-Aminoantipyrine, 98%, C11H13N3O, CAS Number-83-07-8, 4-aminoantipyrene, aminoantipyrin, ampyrone, 4-aminoantipyrine, 4-aminophenazone, metapirazone, solnapyrin-a, aminoazophene, solvapyrin-a, aminoantipyrine, 25g, (5% aq. soln.) clear, 24,273, 100.0, 97.5, CHEBI:59026, Brown-Yellow to Yellow
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
500 g
Packaging:
View all
Plastic Bottle
Catalog number 103150025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-amino-1,5-dimethyl-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Molecular FormulaC11H13N3O
InChI KeyRLFWWDJHLFCNIJ-UHFFFAOYSA-N
SMILESCN1N(C(=O)C(N)=C1C)C1=CC=CC=C1
Molecular Weight (g/mol)203.25
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point105°C to 110°C
Titration with HClO4>=97.5 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only