2-Aminobenzotrifluoride, 99%, Thermo Scientific Chemicals
2-Aminobenzotrifluoride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminobenzotrifluoride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 103181000
Price (USD)/ Each
47.00
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
47.00
Add to cart
2-Aminobenzotrifluoride, 99%, Thermo Scientific Chemicals
Catalog number103181000
Price (USD)/ Each
47.00
-
Add to cart
Chemical Identifiers
CAS88-17-5
IUPAC Name2-(trifluoromethyl)aniline
Molecular FormulaC7H6F3N
InChI KeyVBLXCTYLWZJBKA-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4800 to 1.4830 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only