Thermo Scientific Chemicals

6-Aminocaproic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 103301000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

6-Aminocaproic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 103301000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 103301000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-32-2
IUPAC Name
6-aminohexanoic acid
Molecular Formula
C6H13NO2
InChI Key
SLXKOJJOQWFEFD-UHFFFAOYSA-N
SMILES
NCCCCCC(O)=O
Solubility
(13g/100ml H2O) Clear colorless solution
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
204°C to 209°C (Dec)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text