1x1 image pixel for data collection
6-Aminocaproic acid, 99+%
6-Aminocaproic acid, 99+%
6-Aminocaproic acid, 99+%
6-Aminocaproic acid, 99+%
Thermo Scientific Chemicals

6-Aminocaproic acid, 99+%

CAS: 60-32-2 | C6H13NO2 | 131.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
500 g
10 kg
Packaging:
View all
Plastic bottle
Catalog number 103305000
Price (USD)
285.00
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
285.00
Each
Add to cart
Chemical Identifiers
CAS60-32-2
IUPAC Name6-aminohexanoic acid
Molecular FormulaC6H13NO2
InChI KeySLXKOJJOQWFEFD-UHFFFAOYSA-N
SMILESNCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(13g/100ml H2O) Clear colorless solution
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point204°C to 209°C (Dec)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link