2-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 103320250
Price (USD)/ Each
51.50
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
51.50
Add to cart
2-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Catalog number103320250
Price (USD)/ Each
51.50
-
Add to cart
Chemical Identifiers
CAS89-77-0
IUPAC Name2-amino-4-chlorobenzoic acid
Molecular FormulaC7H6ClNO2
InChI KeyJYYLQSCZISREGY-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to light brown
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Form)Powder
Melting point231°C to 235°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only