4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%, Thermo Scientific Chemicals
4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 103390250
Price (USD)/ Each
398.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
398.00
Add to cart
4-Amino-6-chloro-2-methylmercaptopyrimidine, 97%, Thermo Scientific Chemicals
Catalog number103390250
Price (USD)/ Each
398.00
-
Add to cart
Chemical Identifiers
CAS1005-38-5
IUPAC Name6-chloro-2-(methylsulfanyl)pyrimidin-4-amine
Molecular FormulaC5H6ClN3S
InChI KeyISUXMAHVLFRZQU-UHFFFAOYSA-N
SMILESCSC1=NC(N)=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to beige
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder
Melting point130°C to 134°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only