2-Amino-5-chloropyridine, 98%, Thermo Scientific Chemicals
2-Amino-5-chloropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-chloropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number 103411000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-chloropyridin-2-amine
Molecular FormulaC5H5ClN2
InChI KeyMAXBVGJEFDMHNV-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=N1
Molecular Weight (g/mol)128.56
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Crystalline powder
Melting point134°C to 138°C
Appearance (Color)Beige to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only